Merck
Etiology and incidence of pediatric gallbladder disease.
Surgery (2013-10-01)
Sarah K Walker, Alexandra C Maki, Robert M Cannon, David S Foley, Kristina M Wilson, Laura A Galganski, Chad A Wiesenauer, Sheldon J Bond
ABSTRACT
The spectrum of pediatric biliary tract disease is changing. The goal of this study was to examine the causes and comorbidities of pediatric gallbladder disease at our institution. We performed a retrospective chart review on consecutive patient at Kosair Children's Hospital who underwent cholecystectomy over a 9-year time period ending in 2012. Among the 453 patients in the study group, the average age was 13.3 years and 67.2% were female. Indications for cholecystectomy were gallstones in 285 (63%) and biliary dyskinesia in 140 (33%). Of the patients with gallstones, 68 children (15%) had hemolytic disease. Although the number of cholecystectomies for hemolytic disease was relatively stable throughout our study, the number for biliary dyskinesia and non-hemolytic (cholesterol) cholelithiasis rose by 63% and 216%, respectively. Average body mass index (BMI) for patients with non-hemolytic (cholesterol) stones and biliary dyskinesia were significantly greater than the average BMI for patients with hemolytic stones (P < .0001). In addition, the average BMI for children with non-hemolytic (cholesterol) stones was greater than the average BMI with biliary dyskinesia (P < .0001). Symptomatic gallbladder disease increased over the study period. Biliary dyskinesia and children with non-hemolytic disease are responsible for this increase.
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)