Merck
  • Effects of selected metal oxide nanoparticles on multiple biomarkers in Carassius auratus.
Effects of selected metal oxide nanoparticles on multiple biomarkers in Carassius auratus.
Biomedical and environmental sciences : BES (2013-10-09)
Jun Xia, Hai Zhou Zhao, Guang Hua Lu
ABSTRACT
To study the biological effects of nanoscale copper oxide (nCuO), zinc oxide (nZnO), cerium dioxide (nCeO2) and their mixtures on Carassius auratus. Juvenile fish (Carassius auratus) were exposed to aqueous suspensions of nCuO, nZnO, and nCeO2 (alone and in mixtures) at concentrations of 20, 40, 80, 160, and 320 mg/L. The biomarkers-acetylcholinesterase (AChE) in brain, sodium/potassium-activated ATPase (Na+/K+-ATPase) in gill, and superoxide dismutase (SOD) and catalase (CAT) in liver-were determined after 4 days of exposure. Integrated biomarker response (IBR) was calculated by combining multiple biomarkers into a single value. AChE and SOD activities were significantly inhibited by all test metal oxide nanoparticles (NPs) at high concentrations (⋝160 mg/L) with the exception of nCeO2. Na+/K+-ATPase induction exhibited bell-shaped concentration-response curves. CAT activity was significantly inhibited at concentrations equal to or higher than 160 mg/L. The order of IBR values was nCeO2 ≈ nZnO/nCeO2 ≈ nCuO/nCeO2 < nCuO/nZnO/nCeO2 < nZnO < nCuO < nCuO/nZnO. The joint effect seemed to be synergistic for nCuO/nZnO mixtures, additive for the ternary mixture and less than additive or antagonistic for the binary mixtures containing nCeO2. Concentration-dependent changes of enzymatic activities (AChE, Na+/K+-ATPase, SOD, and CAT) were observed in fish exposed to nanoscale metal oxides. IBR analysis allowed good discrimination between the different exposures and might be a useful tool for the quantification of integrated negative effects induced by NPs toward fish.
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Zinc oxide, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis