Merck
  • Thiol-blocking electrophiles interfere with labeling and detection of protein sulfenic acids.
Thiol-blocking electrophiles interfere with labeling and detection of protein sulfenic acids.
The FEBS journal (2013-10-10)
Julie A Reisz, Erika Bechtold, S Bruce King, Leslie B Poole, Cristina M Furdui
ABSTRACT
Cellular exposure to reactive oxygen species induces rapid oxidation of DNA, proteins, lipids and other biomolecules. At the proteome level, cysteine thiol oxidation is a prominent post-translational process that is implicated in normal physiology and numerous pathologies. Methods for investigating protein oxidation include direct labeling with selective chemical probes and indirect tag-switch techniques. Common to both approaches is chemical blocking of free thiols using reactive electrophiles to prevent post-lysis oxidation or other thiol-mediated cross-reactions. These reagents are used in large excess, and their reactivity with cysteine sulfenic acid, a critical oxoform in numerous proteins, has not been investigated. Here we report the reactivity of three thiol-blocking electrophiles, iodoacetamide, N-ethylmaleimide and methyl methanethiosulfonate, with protein sulfenic acid and dimedone, the structural core of many sulfenic acid probes. We demonstrate that covalent cysteine -SOR (product) species are partially or fully susceptible to reduction by dithiothreitol, tris(2-carboxyethyl)phosphine and ascorbate, regenerating protein thiols, or, in the case of ascorbate, more highly oxidized species. The implications of this reactivity on detection methods for protein sulfenic acids and S-nitrosothiols are discussed.
Sigma-Aldrich
5,5-Dimethyl-1,3-cyclohexanedione, 95%
Supelco
5,5-Dimethyl-1,3-cyclohexanedione, derivatization grade (HPLC), for the determination of aldehyde formaldehyde, ≥99.0%
Sigma-Aldrich
Methyl methanesulfonate, 99%
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
S-Methyl methanethiosulfonate, purum, ≥98.0% (GC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
S-Methyl methanethiosulfonate, 97%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)