Merck
  • Antilisterial activity of lactose monolaurate in milk, drinkable yogurt and cottage cheese.
Antilisterial activity of lactose monolaurate in milk, drinkable yogurt and cottage cheese.
Letters in applied microbiology (2013-10-15)
Y Chen, B Nummer, M K Walsh
ABSTRACT
Lactose monolaurate (LML) was previously found to be an antimicrobial against Listeria monocytogenes in culture medium at concentrations between 3 and 5 mg ml(-1) . In this study, the microbial inhibitory activity of LML in dairy products inoculated with a 5-strain cocktail of clinical isolates of L. monocytogenes was investigated. Addition of LML at a concentration of 5 mg ml(-1) resulted in 4.4, 4.0 and 4.2 log reductions in 0.5% fat, 1% fat and 3.25% fat milks, respectively; 4.1, 4.4, and 3.5 log reductions in nonfat, 1% fat, and 1.5% fat yogurts, respectively; and 4.0 log reductions in both nonfat and 2% fat cottage cheese. The inhibitory effect of LML was only observed at 37°C and not 5°C. Experiments suggest that both the lauric acid and the esterified lactose moiety of LML play roles in the growth inhibition. A novel sugar ester, lactose monolaurate, inhibited the growth of a five-strain cocktail of Listeria monocytogenes in milk, yogurt and cottage cheese. This is the first report of the use of a sugar ester to inhibit the growth of Listeria in food systems.
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, 98%
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)