Merck
  • The complete mitochondrial genome sequence of Sogatella furcifera (Horváth) and a comparative mitogenomic analysis of three predominant rice planthoppers.
The complete mitochondrial genome sequence of Sogatella furcifera (Horváth) and a comparative mitogenomic analysis of three predominant rice planthoppers.
Gene (2013-10-15)
Kai-Jun Zhang, Wen-Chao Zhu, Xia Rong, Jing Liu, Xiu-Lei Ding, Xiao-Yue Hong
ABSTRACT
The white-backed planthopper (WBPH), Sogatella furcifera (Horváth), is one of the most destructive pests of rice crops in many Asian countries. Using long-PCR and shotgun library methods, we sequenced the entire mitochondrial genomes (mt-genomes) of two WBPH individuals. Total lengths of the mt-genome of the two WBPH individuals were 16,612 bp and 16,654 bp with an identical AT content of 76.19%. Among the 13 protein coding genes (PCGs), only nad5 used an atypical initiation codon GTG. Most of the tRNA genes had the typical cloverleaf secondary structure except that the dihydrouridine (DHU) arms in two trnS genes and the TΨC arm of trnG gene did not form a stable stem-loop structure. Similar to the brown planthopper (BPH), Nilaparvata lugens (Stål), and the small brown planthopper (SBPH), Laodelphax striatellus (Fallén), some extraordinary features were observed in the WBPH mt-genome. These include similar gene rearrangement pattern, unusually short length of the atp8 gene and variable numbers of tandem repeat (VNTR) structure in control region. Interestingly, the same tandem repeat unit with stable secondary structure appeared in two different planthoppers, WBPH and SBPH, which belong to two different genera of the Delphacidae. This peculiar feature provides a direct evidence for the close relationship between the two planthoppers and updates our understanding of the evolutionary characteristics of mitochondrial control region. Comparison with two other predominant rice planthoppers (BPH and SBPH) revealed that different PCGs of mitochondria exhibit different evolutionary patterns.
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder