Merck
  • Apoptosis-like death in Leishmania donovani promastigotes induced by eugenol-rich oil of Syzygium aromaticum.
Apoptosis-like death in Leishmania donovani promastigotes induced by eugenol-rich oil of Syzygium aromaticum.
Journal of medical microbiology (2013-10-29)
Mohammad Islamuddin, Dinkar Sahal, Farhat Afrin
ABSTRACT
Leishmaniasis consists of a complex spectrum of infectious diseases with worldwide distribution of which visceral leishmaniasis or kala-azar caused by Leishmania donovani is the most devastating. In the absence of vaccines, chemotherapy remains the mainstay for the control of leishmaniasis. The drugs of choice are expensive and associated with multiple adverse side effects. Because of these limitations, the development of new antileishmanial compounds is imperative and plants offer prospects in this regard. The present work was conducted to study the antileishmanial potential of oil from Syzygium aromaticum flower buds (clove). The S. aromaticum oil was characterized by gas chromatography and GC-MS and eugenol as well as eugenyl acetate were found to be the most abundant compounds, composing 59.75 % and 29.24 %, respectively of the oil. Our findings have shown that eugenol-rich essential oil from S. aromaticum (EROSA) possesses significant activity against L. donovani, with 50 % inhibitory concentration of 21 ± 0.16 µg ml(-1) and 15.24 ± 0.14 µg ml(-1), respectively, against promastigotes and intracellular amastigotes. Alterations in cellular morphology and growth reversibility assay substantiated the leishmanicidal activity of EROSA. The leishmanicidal effect was mediated via apoptosis as confirmed by externalization of phosphatidylserine, DNA nicking by TdT-mediated dUTP nick-end labelling (TUNEL) assay, dyskinetoplastidy, cell cycle arrest at sub-G0-G1 phase, loss of mitochondrial membrane potential and reactive oxygen species generation. EROSA presented no adverse cytotoxic effects against murine macrophages even at 200 µg ml(-1). Our studies authenticate the promising antileishmanial activity of EROSA, which is mediated by programmed cell death, and, accordingly, EROSA may be a source of novel agents for the treatment of leishmaniasis.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture