Merck
  • Negative production of acetoin in the photochemistry of aqueous pyruvic acid.
Negative production of acetoin in the photochemistry of aqueous pyruvic acid.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-31)
Alexis J Eugene, Sha-Sha Xia, Marcelo I Guzman
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Pyruvic acid, 95%