Merck
  • Pain and intramuscular release of algesic substances in the masseter muscle after experimental tooth-clenching exercises in healthy subjects.
Pain and intramuscular release of algesic substances in the masseter muscle after experimental tooth-clenching exercises in healthy subjects.
Journal of orofacial pain (2013-10-31)
Andreas Dawson, Bijar Ghafouri, Björn Gerdle, Thomas List, Peter Svensson, Malin Ernberg
ABSTRACT
To investigate whether experimental tooth clenching leads to a release of algesic substances in the masseter muscle. Thirty healthy subjects (16 females, 14 males) participated. During two sessions, separated by at least 1 week, intramuscular microdialysis was performed to collect masseter muscle 5-hydroxytryptamine (5-HT) and glutamate as well as the metabolic markers pyruvate and lactate. Two hours after the start of microdialysis, participants were randomized to a 20-min repetitive experimental tooth-clenching task (50% of maximal voluntary contraction) or a control session (no clenching). Pain and fatigue were measured throughout. The Friedman and Wilcoxon tests were used for statistical analyses. No alterations were observed in the concentrations of 5-HT, glutamate, pyruvate, and lactate over time in the clenching or control session, or between sessions at various time points. Pain (P < .01) and fatigue (P < .01) increased significantly over time in the clenching session and were significantly higher after clenching than in the control session (P < .01). Low levels of pain and fatigue developed with this experimental tooth-clenching model, but they were not associated with an altered release of 5-HT, glutamate, lactate, or pyruvate. More research is required to elucidate the peripheral release of algesic substances in response to tooth clenching.
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG