Merck
  • The sarcoplasmic-endoplasmic reticulum Ca(2+)-ATPase (SERCA) is the likely molecular target for the acute toxicity of the brominated flame retardant hexabromocyclododecane (HBCD).
The sarcoplasmic-endoplasmic reticulum Ca(2+)-ATPase (SERCA) is the likely molecular target for the acute toxicity of the brominated flame retardant hexabromocyclododecane (HBCD).
Chemico-biological interactions (2013-11-06)
Fawaz Al-Mousa, Francesco Michelangeli
ABSTRACT
Hexabromocyclododecane (HBCD) is a widely utilised brominated flame retardant (BFR). It has been shown to bio-accumulate within organisms, including man, and possibly cause neurological disorders. The acute neurotoxicity of HBCD, and six other unrelated BFRs, were assessed in SH-SY5Y human neuroblastoma cells by 24h viability assays and HBCD proved to be the most lethal (LC50, 3μM). In addition, the effects of these BFRs were also assessed for their potency at inhibiting the sarcoplasmic-endoplasmic reticulum Ca(2+) ATPase (SERCA) derived from the SH-SY5Y cells and again HBCD was the most potent (IC50, 2.7μM). The data for the other BFRs tested showed a direct correlation (coefficient 0.94) between the potencies of inducing cell death and inhibiting the Ca(2+) ATPase, indicating that SERCA is likely to be the molecular target for acute toxicity. Mechanistic studies of HBCD on the Ca(2+) ATPase suggest that it affects ATP binding, phosphorylation as well as the E2 to E1 transition step.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%