Merck
Sodium fluoride inhibits MMP-2 and MMP-9.
Journal of dental research (2013-11-08)
M T Kato, A Bolanho, B L Zarella, T Salo, L Tjäderhane, M A R Buzalaf
ABSTRACT
The importance of fluoride (F) in preventing dental caries by favorably interfering in the demineralization-remineralization processes is well-established, but its ability to inhibit matrix metalloproteinases (MMPs), which could also help to prevent dentin caries, has not been investigated. This study assessed the ability of F to inhibit salivary and purified human gelatinases MMPs-2 and -9. Saliva was collected from 10 healthy individuals. Pooled saliva was centrifuged, and supernatants were incubated for 1 hr at 37°C and subjected to zymography. Sodium fluoride (50-275 ppm F) was added to the incubation buffer. The reversibility of the inhibition of MMPs-2 and -9 by NaF was tested by the addition of NaF (250-5,000 ppm F) to the incubation buffer, after which an additional incubation was performed in the absence of F. F decreased the activities of pro- and active forms of salivary and purified human MMPs in a dose-response manner. Purified gelatinases were completely inhibited by 200 ppm F (IC50 = 100 and 75 ppm F for MMPs-2 and -9, respectively), and salivary MMP-9 by 275 ppm F (IC50 = 200 ppm F). Inhibition was partially reversible at 250-1,500 ppm F, but was irreversible at 5,000 ppm F. This is the first study to describe the ability of NaF to inhibit MMPs completely.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis