Merck
  • Selectivity of binding of PEGs and PEG-like oligomers to anti-PEG antibodies induced by methoxyPEG-proteins.
Selectivity of binding of PEGs and PEG-like oligomers to anti-PEG antibodies induced by methoxyPEG-proteins.
Molecular immunology (2013-11-10)
Mark G P Saifer, L David Williams, Monika A Sobczyk, Shawnya J Michaels, Merry R Sherman
ABSTRACT
The use of methoxypoly(ethylene glycol) (mPEG) in PEG conjugates of proteins and non-protein therapeutic agents has led to the recognition that the polymer components of such conjugates can induce anti-PEG antibodies (anti-PEGs) that may accelerate the clearance and reduce the efficacy of the conjugates. Others have classified anti-PEGs as "methoxy-specific" or "backbone-specific". The results of our previous research on anti-PEGs in the sera of rabbits immunized with mPEG or hydroxyPEG (HO-PEG) conjugates of three unrelated proteins were consistent with that classification (Sherman, M.R., et al., 2012. Bioconjug. Chem. 23, 485-499). Enzyme-linked immunosorbent assays (ELISAs) were performed on rabbit antisera and rabbit monoclonal anti-PEGs with competitors including 10 kDa mPEG, 10 kDa PEG diol and six linear or cyclic oligomers of oxyethylene (CH2CH2O), with molecular weights of ca. 150-264 Da. Our results demonstrate that (1) the binding affinities of anti-mPEGs depend more on the backbone lengths of the polymers and the hydrophobicities of their end-groups than on their resemblance to the methoxy terminus of the immunogenic polymer; (2) anti-PEGs raised against HO-PEG-proteins are not directed against the terminal hydroxy group, but against the backbone; (3) rabbit anti-PEGs bind to and distinguish among PEG-like oligomers with as few as three oxyethylene groups; and (4) none of the monoclonal or polyclonal anti-PEGs was absolutely "methoxy-specific" or "backbone-specific", but displayed distinct relative selectivities. If these results are relevant to human immune responses, the clinical use of stable conjugates of HO-PEG with proteins and non-protein therapeutic agents would be expected to produce fewer and less intense immune responses than those induced by conjugates with mPEG or PEGs with larger alkoxy groups.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350