Merck
  • Transient kinetics and rate-limiting steps for the processive cellobiohydrolase Cel7A: effects of substrate structure and carbohydrate binding domain.
Transient kinetics and rate-limiting steps for the processive cellobiohydrolase Cel7A: effects of substrate structure and carbohydrate binding domain.
Biochemistry (2013-11-16)
Nicolaj Cruys-Bagger, Hirosuke Tatsumi, Guilin Robin Ren, Kim Borch, Peter Westh
ABSTRACT
Cellobiohydrolases are exoacting, processive enzymes that effectively hydrolyze crystalline cellulose. They have attracted considerable interest because of their role in both natural carbon cycling and industrial enzyme cocktails used for the deconstruction of cellulosic biomass, but many mechanistic and regulatory aspects of their heterogeneous catalysis remain poorly understood. Here, we address this by applying a deterministic model to real-time kinetic data with high temporal resolution. We used two variants of the cellobiohydrolase Cel7A from Hypocrea jecorina , and three types of cellulose as substrate. Analysis of the pre-steady-state regime allowed delineation rate constants for both fast and slow steps in the enzymatic cycle and assessment of how these constants influenced the rate of hydrolysis at quasi-steady state. Processive movement on the cellulose strand advanced with characteristic times of 0.15-0.7 s per step at 25 °C, and the rate was highest on amorphous substrate. The cellulose binding module was found to raise this rate on crystalline, but not on amorphous, substrate. The rapid processive movement signified high intrinsic reactivity, but this parameter had marginal influence on the steady-state rate. This was because dissociation and association were slower and, hence, rate limiting. Specifically, the dissociation from the strand was found to occur with characteristic times of 45-100 s. This meant that dissociation was the bottleneck, except at very low substrate loads (0.5-1 g/L), where association became slower.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Cellobiohydrolase I from Hypocrea jecorina, 100 U/mg, recombinant, expressed in corn
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)