Merck
Gold nanosponges: green synthesis, characterization, and cytotoxicity.
Journal of nanoscience and nanotechnology (2013-11-20)
Yang-Wei Lin, Yi-Cheng Chen, Chia-Wei Wang, Wen-Tsen Chen, Chao-Ming Liu, Chung-Yu Chen, Huan-Tsung Chang
ABSTRACT
A simply approach for the synthesis of Au nanostructures in tea infusions at room temperature is developed. By controlling the concentrations of tea infusions, Au nanostructures in various shapes and sizes have been prepared. From 1 x (original concentration) and 0.01 x (100 times diluted) tea infusions, 52.2 +/- 8.1 nm Au nanosponges (T-Au NSs) and 23 +/- 2 nm spherical Au nanoparticles (T-Au NPs) were prepared. The phytochemicals present on the surface of T-Au NSs were proved by surface-assisted laser desorption/ionization mass spectrometry, Fourier transform infrared spectrometry and capillary electrophoresis coupled with UV detection. The energy-dispersive X-ray spectroscopy and powder X-ray diffraction data reveal pure crystalline structures of the T-Au NSs. The dark field scattering images observe that the T-Au NSs have significant affinity toward HeLa cells. The cytotoxicity of T-Au NSs on HeLa cells is through caspase-3 activation.
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis