Merck
  • Protective action of n-3 fatty acids on benzo[a]pyrene-induced apoptosis through the plasma membrane remodeling-dependent NHE1 pathway.
Protective action of n-3 fatty acids on benzo[a]pyrene-induced apoptosis through the plasma membrane remodeling-dependent NHE1 pathway.
Chemico-biological interactions (2013-11-20)
Béatrice Dendelé, Xavier Tekpli, Kévin Hardonnière, Jørn A Holme, Laure Debure, Daniel Catheline, Volker M Arlt, Eszter Nagy, David H Phillips, Steinar Ovrebø, Steen Mollerup, Mallory Poët, Martine Chevanne, Vincent Rioux, Marie-Thérèse Dimanche-Boitrel, Odile Sergent, Dominique Lagadic-Gossmann
ABSTRACT
Plasma membrane is an early target of polycyclic aromatic hydrocarbons (PAH). We previously showed that the PAH prototype, benzo[a]pyrene (B[a]P), triggers apoptosis via DNA damage-induced p53 activation (genotoxic pathway) and via remodeling of the membrane cholesterol-rich microdomains called lipid rafts, leading to changes in pH homeostasis (non-genotoxic pathway). As omega-3 (n-3) fatty acids can affect membrane composition and function or hamper in vivo PAH genotoxicity, we hypothesized that addition of physiologically relevant levels of polyunsaturated n-3 fatty acids (PUFAs) might interfere with B[a]P-induced toxicity. The effects of two major PUFAs, docosahexaenoic acid (DHA) and eicosapentaenoic acid (EPA), were tested on B[a]P cytotoxicity in the liver epithelial cell line F258. Both PUFAs reduced B[a]P-induced apoptosis. Surprisingly, pre-treatment with DHA increased the formation of reactive B[a]P metabolites, resulting in higher levels of B[a]P-DNA adducts. EPA had no apparent effect on B[a]P metabolism or related DNA damage. EPA and DHA prevented B[a]P-induced apoptotic alkalinization by affecting Na(+)/H(+) exchanger 1 activity. Thus, the inhibitory effects of omega-3 fatty acids on B[a]P-induced apoptosis involve a non-genotoxic pathway associated with plasma membrane remodeling. Our results suggest that dietary omega-3 fatty acids may have marked effects on the biological consequences of PAH exposure.
Product Description
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland