Merck
  • Intrathecal ultra-low dose naloxone enhances the antihyperalgesic effects of morphine and attenuates tumor necrosis factor-α and tumor necrosis factor-α receptor 1 expression in the dorsal horn of rats with partial sciatic nerve transection.
Intrathecal ultra-low dose naloxone enhances the antihyperalgesic effects of morphine and attenuates tumor necrosis factor-α and tumor necrosis factor-α receptor 1 expression in the dorsal horn of rats with partial sciatic nerve transection.
Anesthesia and analgesia (2013-11-22)
Chih-Ping Yang, Chen-Hwan Cherng, Ching-Tang Wu, Hui-Yi Huang, Pao-Luh Tao, Sing-Ong Lee, Chih-Shung Wong
ABSTRACT
Glutamate homeostasis and microglia activation play an important role in the development and maintenance of neuropathic pain. We designed this investigation to examine whether ultra-low dose naloxone administered alone or in combination with morphine could alter the concentration of the excitatory amino acids (EAAs) glutamate and aspartate, as well as the expression of tumor necrosis factor-α (TNF-α) and its receptors (TNFR1 and TNFR2) in the spinal cord dorsal horn of rats with partial sciatic nerve transection (PST). Male Wistar rats underwent intrathecal catheter implantation for drug delivery and were divided in 7 groups: sham-operated + saline (sham), PST + saline (S), PST + 15 ng naloxone (n), PST + 15 µg naloxone (N), PST + 10 µg morphine (M), PST + 15 ng naloxone + 10 µg morphine (Mn), PST + 15 µg naloxone + 10 µg morphine (MN). Thermal withdrawal latency and mechanical withdrawal threshold, TNF-α and TNFR expression in the spinal cord and dorsal root ganglia, and EAAs glutamate and aspartate concentration in cerebrospinal fluid dialysates were measured. Ten days after PST, rats developed hyperalgesia (P < 0.0001) and allodynia (P < 0.0001), and increased TNF-α (P < 0.0001) and TNFR1 expression (P = 0.0009) were measured in the ipsilateral spinal cord dorsal horn. The antihyperalgesic and antiallodynic effects of morphine (10 μg) were abolished by high-dose naloxone (15 μg; P = 0.0031) but enhanced by ultra-low dose naloxone (15 ng; P = 0.0015), and this was associated with a reduction of TNF-α (P < 0.0001) and TNFR1 (P = 0.0009) expression in the spinal cord dorsal horn and EAAs concentration (glutamate: P = 0.0001; aspartate: P = 0.004) in cerebrospinal fluid dialysate. Analysis of variance (ANOVA) or Student t test with Bonferroni correction were used for statistical analysis. Ultra-low dose naloxone enhances the antihyperalgesia and antiallodynia effects of morphine in PST rats, possibly by reducing TNF-α and TNFR1 expression, and EAAs concentrations in the spinal dorsal horn. Ultra-low dose naloxone may be a useful adjuvant for increasing the analgesic effect of morphine in neuropathic pain conditions.
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material