Merck
  • Melamine nanosensing with chondroitin sulfate-reduced gold nanoparticles.
Melamine nanosensing with chondroitin sulfate-reduced gold nanoparticles.
Journal of nanoscience and nanotechnology (2013-11-26)
Hwa Jung Noh, Hyun-Seok Kim, Seonho Cho, Youmie Park
ABSTRACT
Gold nanoparticles were green-synthesized using a glycosaminoglycan, chondroitin sulfate, as the reducing agent by mixing Au3+ and chondroitin sulfate under heating. Chondroitin sulfate-reduced gold nanoparticles were characterized by UV-Vis spectrophotometry, high resolution transmission electron microscopy and atomic force microscopy. The yield of Au3+ to Au0 was measured as 80.1% by inductively coupled plasma-atomic emission spectroscopy. A mostly spherical shape, with an average diameter of 44.68 +/- 11.25 nm, was observed from the atomic force microscopy images. Using chondroitin sulfate-reduced gold nanoparticles, we developed a melamine nanosensor that provides a simplified method to detect melamine in infant formula. With an increase in the melamine concentration in the gold nanoparticle solution, the characteristic surface plasmon resonance band of gold nanoparticles at 530 nm decreased, whereas a new peak appeared at 620 nm. There was a linear relationship between the absorbance ratio (A620/A530) and the melamine concentration in the range of 0.1-10 microM. The practical use of the proposed method was verified by quantifying melamine spiked in real infant formula at concentrations as low as 12.6 ppb. The nanosensing of melamine using chondroitin sulfate-reduced gold nanoparticles can be a promising technique for quick on-site melamine screening of milk products.
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Supelco
Melamine, analytical standard
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Melamine, 99%
Supelco
Melamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, layer thickness 100 Å, 99.999% (Au), L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Chondroitin 4-sulfate sodium salt from bovine trachea, mixture of isomers, main component: chondroitin 4-sulfate
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis