Merck
  • Skeletal muscle glycogen phosphorylase is irreversibly inhibited by mercury: molecular, cellular and kinetic aspects.
Skeletal muscle glycogen phosphorylase is irreversibly inhibited by mercury: molecular, cellular and kinetic aspects.
FEBS letters (2013-11-26)
Ximing Xu, Cécile Mathieu, Solène Emmanuelle Boitard, Julien Dairou, Jean-Marie Dupret, Onnik Agbulut, Fernando Rodrigues-Lima
ABSTRACT
Muscle glycogen phosphorylase (GP) plays an important role in muscle functions. Mercury has toxic effects in skeletal muscle leading to muscle weakness or cramps. However, the mechanisms underlying these toxic effects are poorly understood. We report that GP is irreversibly inhibited by inorganic (Hg(2+)) and organic (CH3Hg(+)) mercury (IC50=380 nM and kinact=600 M(-1) s(-1) for Hg(2+) and IC50=43 μM and kinact=13 M(-1) s(-1) for CH3Hg(+)) through reaction of these compounds with cysteine residues of the enzyme. Our data suggest that the irreversible inhibition of GP could represent one of the mechanisms that contribute to mercury-dependent muscle toxicity.
Product Description
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis