Merck
  • De novo protein crystal structure determination from X-ray free-electron laser data.
De novo protein crystal structure determination from X-ray free-electron laser data.
Nature (2013-11-26)
Thomas R M Barends, Lutz Foucar, Sabine Botha, R Bruce Doak, Robert L Shoeman, Karol Nass, Jason E Koglin, Garth J Williams, Sébastien Boutet, Marc Messerschmidt, Ilme Schlichting
ABSTRACT
The determination of protein crystal structures is hampered by the need for macroscopic crystals. X-ray free-electron lasers (FELs) provide extremely intense pulses of femtosecond duration, which allow data collection from nanometre- to micrometre-sized crystals in a 'diffraction-before-destruction' approach. So far, all protein structure determinations carried out using FELs have been based on previous knowledge of related, known structures. Here we show that X-ray FEL data can be used for de novo protein structure determination, that is, without previous knowledge about the structure. Using the emerging technique of serial femtosecond crystallography, we performed single-wavelength anomalous scattering measurements on microcrystals of the well-established model system lysozyme, in complex with a lanthanide compound. Using Monte-Carlo integration, we obtained high-quality diffraction intensities from which experimental phases could be determined, resulting in an experimental electron density map good enough for automated building of the protein structure. This demonstrates the feasibility of determining novel protein structures using FELs. We anticipate that serial femtosecond crystallography will become an important tool for the structure determination of proteins that are difficult to crystallize, such as membrane proteins.
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein