Merck
  • Protection of corneal epithelial stem cells prevents ultraviolet A damage during corneal collagen cross-linking treatment for keratoconus.
Protection of corneal epithelial stem cells prevents ultraviolet A damage during corneal collagen cross-linking treatment for keratoconus.
The British journal of ophthalmology (2013-11-30)
Jonathan E Moore, Sarah D Atkinson, Dimitri T Azar, Jenny Worthington, C Stephen Downes, David G Courtney, C B Tara Moore
ABSTRACT
Cross-linking of the cornea is usually carried out at a young age as a treatment to manage ectasia. The corneal limbal region contains delicate long-lived stem cells, which could potentially be deleteriously affected by Ultraviolet A (UV-A) radiation. Damage to these stem cells may not demonstrate as a clinical problem for many years subsequent to cross-linking treatment. UV-A radiation is known to have potential mutagenic effects upon mammalian DNA and can result in cancer. Cultured corneal epithelial cells and ex vivo corneal tissue were treated with the standard clinical cross-linking protocol for UV-A irradiation. 8-hydroxydeoxyguansoine (8-OHdG) and cyclin-dependent kinase inhibitor genes (CDKN1A and CDKN2A) were assayed as markers of DNA damage using immunohistochemistry, ELISA and quantitative real time PCR. Staining of treated limbal tissue demonstrated the presence of 8-OHdG within p63 positive basal limbal cells. Levels of 8-OHdG and CDKN1A mRNA were found to be significantly increased in cultured corneal epithelial cells and limbal epithelial cells but no increase was demonstrated with the use of a polymethyl methylacrylate protective cover. This study provides evidence that oxidative nuclear DNA damage can occur through cross-linking in layers of corneal epithelial cells at the limbus and that this can be easily prevented by covering the limbus.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture