Merck
  • Naproxen intercalates with DNA and causes photocleavage through ROS generation.
Naproxen intercalates with DNA and causes photocleavage through ROS generation.
The FEBS journal (2013-12-05)
Mohammed A Husain, Zahid Yaseen, Sayeed U Rehman, Tarique Sarwar, Mohammad Tabish
ABSTRACT
Naproxen is an important non-steroidal anti-inflammatory drug with many pharmacological and biological properties. In this study, we have attempted to ascertain the mode of action and the mechanism of binding of naproxen to DNA. We have also demonstrated that, upon irradiation with white light, naproxen generates reactive oxygen species, causing DNA cleavage. Generation of reactive oxygen species from photo-irradiated naproxen as determined spectrophotometrically was found to lead to nicking of plasmid DNA as analyzed by agarose gel electrophoresis. Without photo-irradiation, naproxen binds to DNA and forms drug-DNA complexes as revealed by spectroscopic techniques. Several experiments such as determination of the effect of urea, iodide-induced quenching and a competitive binding assay with ethidium bromide showed that naproxen binds to DNA primarily in an intercalative manner. These observations were further supported by CD analysis, viscosity measurements and molecular docking. Using DNA as a template, fluorescence resonance energy transfer between naproxen and ethidium bromide was also observed, further strengthening the evidence for intercalative binding of naproxen with DNA.
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Genomic, unsheared
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Gold etchant, standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, VETRANAL®, analytical standard