Merck
  • Involvement of CTR1 and ATP7A in lead (Pb)-induced copper (Cu) accumulation in choroidal epithelial cells.
Involvement of CTR1 and ATP7A in lead (Pb)-induced copper (Cu) accumulation in choroidal epithelial cells.
Toxicology letters (2013-12-10)
Gang Zheng, Jieqiong Zhang, Yan Xu, Xuefeng Shen, Han Song, Jinfei Jing, Wenjing Luo, Wei Zheng, Jingyuan Chen
ABSTRACT
The blood-cerebrospinal fluid barrier (BCB) plays a key role in maintaining copper (Cu) homeostasis in the brain. Cumulative evidences indicate that lead (Pb) exposure alters cerebral Cu homeostasis, which may underlie the development of neurodegenerative diseases. This study investigated the roles of Cu transporter 1 (CTR1) and ATP7A, two Cu transporters, in Pb-induced Cu accumulation in the choroidal epithelial cells. Pb exposure resulted in increased intracellular (64)Cu retention, accompanying with up-regulated CTR1 level. Knockdown of CTR1 using siRNA before Pb exposure diminished the Pb-induced increase of (64)Cu uptake. The expression level of ATP7A was down-regulated following the Pb exposure. ATP7A siRNA knockdown, or PCMB treatment, inhibited the (64)Cu efflux from the cells, while the following additional incubation with Pb failed to further increase the intracellular (64)Cu retention. Cu exposure, or intracellular Cu accumulation following the tetracycline (Tet)-induced overexpression of CTR1, did not result in significant change in ATP7A expression. Taken together, these data indicate that CTR1 and ATP7A play important roles in Cu transport in choroidal epithelial cells, and the Pb-induced intracellular Cu accumulation appears to be mediated, at least in part, via the alteration of CTR1 and ATP7A expression levels following Pb exposure.
Product Description
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Copper (O), BCR®, certified reference material, rod
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper (S, P), BCR®, certified reference material, chips
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) acetate trihydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Lead(II) acetate trihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%