Merck
  • Effect of a cholesterol-rich lipid environment on the enzymatic activity of reconstituted hyaluronan synthase.
Effect of a cholesterol-rich lipid environment on the enzymatic activity of reconstituted hyaluronan synthase.
Biochemical and biophysical research communications (2013-12-18)
Pawared Ontong, Yasuyo Hatada, Shun'ichiro Taniguchi, Ikuko Kakizaki, Naoki Itano
ABSTRACT
Hyaluronan synthase (HAS) is a unique membrane-associated glycosyltransferase and its activity is lipid dependent. The dependence however is not well understood, especially in vertebrate systems. Here we investigated the functional association of hyaluronan synthesis in a cholesterol-rich membrane-environment. The culture of human dermal fibroblasts in lipoprotein-depleted medium attenuated the synthesis of hyaluronan. The sequestration of cellular cholesterol by methyl-ß-cyclodextrin also decreased the hyaluronan production of fibroblasts, as well as the HAS activity. To directly evaluate the effects of cholesterol on HAS activity, a recombinant human HAS2 protein with a histidine-tag was expressed as a membrane protein by using a baculovirus system, then successfully solubilized, and isolated by affinity chromatography. When the recombinant HAS2 proteins were reconstituted into liposomes composed of both saturated phosphatidylcholine and cholesterol, this provided a higher enzyme activity as compared with the liposomes formed by phosphatidylcholine alone. Cholesterol regulates HAS2 activity in a biphasic manner, depending on the molar ratio of phosphatidylcholine to cholesterol. Furthermore, the activation profiles of different lipid compositions were determined in the presence or absence of cholesterol. Cholesterol had the opposite effect on the HAS2 activity in liposomes composed of phosphatidylethanolamine or phosphatidylserine. Taken together, the present data suggests a clear functional association between HAS activity and cholesterol-dependent alterations in the physical and chemical properties of cell membranes.
Product Description
Sodium hyaluronate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-90,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide