Merck
  • Protective role of JAK/STAT signaling against renal fibrosis in mice with unilateral ureteral obstruction.
Protective role of JAK/STAT signaling against renal fibrosis in mice with unilateral ureteral obstruction.
Clinical immunology (Orlando, Fla.) (2013-12-18)
Kiyomi Koike, Seiji Ueda, Sho-ichi Yamagishi, Hideo Yasukawa, Yusuke Kaida, Miyuki Yokoro, Kei Fukami, Akihiko Yoshimura, Seiya Okuda
ABSTRACT
Inflammation is involved in renal fibrosis, a final common pathway for kidney diseases. To clarify how JAK/STAT/SOCS system was involved in renal fibrosis, UUO was induced in BALB/c or SOCS3(+/-) mice in the presence or absence of JAK inhibitor-incorporated nanoparticle (pyridine6-PGLA). UUO increased pSTAT3 and subsequently elevated SOCS3 levels in the obstructed kidneys. pSTAT3 levels were further increased in SOCS3(+/-) mice. UUO-induced renal fibrosis was markedly suppressed in SOCS3(+/-) mice, while it was aggravated by pre-treatment with pyridine6-PGLA. Although there were no differences in renal mRNA levels of TGF-β and collagens between wild and SOCS3(+/-) mice, MMP-2 activity was enhanced in SOCS3(+/-) UUO mice. Activated MMP-2 was completely suppressed by pyridine6-PGLA-pre-treatment. TNF-α one of JAK/STAT activators, increased pSTAT3 levels and subsequently induced MMP-2 activation in proximal tubular cells. These results suggest that JAK/STAT3 signaling may play a role in repair process of renal fibrosis in UUO partly via MMP-2 activation.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture