Merck
  • Statistical media and process optimization for biotransformation of rice bran to vanillin using Pediococcus acidilactici.
Statistical media and process optimization for biotransformation of rice bran to vanillin using Pediococcus acidilactici.
Indian journal of experimental biology (2014-01-15)
Baljinder Kaur, Debkumar Chakraborty
ABSTRACT
An isolate of P. acidilactici capable of producing vanillin from rice bran was isolated from a milk product. Response Surface Methodology was employed for statistical media and process optimization for production of biovanillin. Statistical medium optimization was done in two steps involving Placket Burman Design and Central Composite Response Designs. The RSM optimized vanillin production medium consisted of 15% (w/v) rice bran, 0.5% (w/v) peptone, 0.1% (w/v) ammonium nitrate, 0.005% (w/v) ferulic acid, 0.005% (w/v) magnesium sulphate, and 0.1% (v/v) tween-80, pH 5.6, at a temperature of 37 degrees C under shaking conditions at 180 rpm. 1.269 g/L vanillin was obtained within 24 h of incubation in optimized culture medium. This is the first report indicating such a high vanillin yield obtained during biotransformation of ferulic acid to vanillin using a Pediococcal isolate.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG