Merck
  • Implant placement in the aesthetic zone following an autogenous bone graft from an intraoral site: a case study.
Implant placement in the aesthetic zone following an autogenous bone graft from an intraoral site: a case study.
Primary dental journal (2014-01-29)
Jerome P Sullivan
ABSTRACT
In the front of the mouth, the treatment of such a bony defect carries the risk of a poor aesthetic outcome. High patient expectations, combined with the difficulty of recreating a large mass of viable bone in which a dental implant will be biomechanically stable is a real challenge. PURPOSE OF TREATMENT: The intention of this treatment was to replace the patient's chronically infected upper right incisor with a dental implant that was both biomechanically and aesthetically stable for the long term. Four months after extraction of the upper right central incisor, under local anaesthetic, a bone block was harvested from the right retromolar area of the patient's mandible and grafted into the defect. This was secured with a single bone screw. Small deficiencies between the block and the host site were filled in using bovine derived particulate bone (Bio Oss, Geistlich Biomaterials, Wolhuser, Germany). A resorbable porcine derived membrane (Bio-Gide, Geistlich Biomaterials, Wolhuser, Germany), held in position by two bone tacks (Frios, Friadent, Mannheim, Germany), was used to stabilse the graft. Following close clinical and radiographic monitoring of graft maturation, the bone screw was removed at three months and one week later the implant placed. A bone level impression was taken at surgery, and the implant left submerged for four months before exposure and abutment connection. After a further period of two months to facilitate soft tissue contouring the definitive crown was cemented. The implant tooth has been in trouble free function for eighteen months. In that period, the marginal bone levels and gingival margin around the implant have remained stable and aesthetically satisfactory.
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder