Merck
  • Remineralization efficacy of a toothpaste containing 8% arginine and calcium carbonate on enamel surface.
Remineralization efficacy of a toothpaste containing 8% arginine and calcium carbonate on enamel surface.
American journal of dentistry (2014-02-01)
Yajing Huang, Yanxia Duan, Yingzi Qian, Rui Huang, Zhengyan Yang, Yueheng Li, Zhi Zhou
ABSTRACT
To investigate the remineralization efficacy of different types of toothpastes on initial enamel lesions in vitro. Artificial initial lesions were created on 150 enamel discs from freshly extracted bovine incisors. These enamel discs were divided into five groups. The test treatment consisted of undiluted Colgate Sensitive Pro-Relief Toothpaste containing 8.0% arginine, calcium carbonate and 1,450 ppm fluoride that was applied on the enamel surface under a pH-cycling including 4 x 3-minute application daily for 12 days and soaked in remineralizing solution during the untreated periods. The two other test products were commercial products: Crest Cavity Protection Toothpaste, containing 0.11% fluoride and GC Tooth Mousse, a professional remineralizing treatment paste (the active ingredients: casein phosphopeptide - amorphous calcium phosphate, fluoride). NaF solution (0.14% fluoride) was used as the positive control, while double distilled water (ddH2O) was used as the negative control. The remineralization of enamel discs was evaluated using Knoop hardness test, confocal laser scanning microscopy (CLSM) and polarized light microscopy (PLM), and the caries lesion depth was quantified using an image analyzer. The data were analyzed by ANOVA. All test products showed a recovery of the Knoop Hardness Number (KHN) after remineralization cycling treatment. The recovery of enamel KHN for Colgate Sensitive Pro-Relief, GC Mousse, Crest toothpasteand NaF groups were 44.53 +/- 6.72%, 35.00 +/- 7.83%, 24.56 +/- 5.95% and 42.51 +/- 6.74% respectively, while the recovery of negative control group was 18.99 +/- 4.98%. PLM results indicated the lesion depth recovery of 49.63 +/- 8.06%, 35.08 +/- 2.19%, 22.60 +/- 7.30% and 53.20 +/- 1.48% respectively, which were also significantly greater than that of the negative group (20.51 +/- 4.80%). CLSM analysis showed a reduction of average area, and total and average dye fluorescence of the lesions after treatment. The Colgate Sensitive Pro-Relief group presented significantly greater remineralization than the other toothpaste groups, while the Crest toothpaste group showed the lowest remineralization ability.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride 0.5 M solution
SAFC
L-Arginine
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis