Merck
  • Differential role of ethylene and hydrogen peroxide in dark-induced stomatal closure.
Differential role of ethylene and hydrogen peroxide in dark-induced stomatal closure.
Pakistan journal of biological sciences : PJBS (2014-02-13)
R K Kar, N Parvin, D Laha
ABSTRACT
Regulation of stomatal aperture is crucial in terrestrial plants for controlling water loss and gaseous exchange with environment. While much is known of signaling for stomatal opening induced by blue light and the role of hormones, little is known about the regulation of stomatal closing in darkness. The present study was aimed to verify their role in stomatal regulation in darkness. Epidermal peelings from the leaves of Commelina benghalensis were incubated in a defined medium in darkness for 1 h followed by a 1 h incubation in different test solutions [H2O2, propyl gallate, ethrel (ethylene), AgNO3, sodium orthovanadate, tetraethyl ammonium chloride, CaCl2, LaCl3, separately and in combination] before stomatal apertures were measured under the microscope. In the dark stomata remained closed under treatments with ethylene and propyl gallate but opened widely in the presence of H2O2 and AgNO3. The opening effect was largely unaffected by supplementing the treatment with Na-vanadate (PM H+ ATPase inhibitor) and tetraethyl ammonium chloride (K(+)-channel inhibitor) except that opening was significantly inhibited by the latter in presence of H2O2. On the other hand, H2O2 could not override the closing effect of ethylene at any concentrations while a marginal opening of stomata was found when Ag NO3 treatment was given together with propyl gallate. CaCl2 treatment opened stomata in the darkness while LaCl3 maintained stomata closed. A combination of LaCl3 and propyl gallate strongly promoted stomatal opening. A probable action of ethylene in closing stomata of Commelina benghalensis in dark has been proposed.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis