Merck
  • Benzene, toluene and xylenes levels in new and used vehicles of the same model.
Benzene, toluene and xylenes levels in new and used vehicles of the same model.
Journal of environmental sciences (China) (2014-02-21)
Joanna Faber, Krzysztof Brodzik, Anna Golda-Kopek, Damian Lomankiewicz
ABSTRACT
The aim of this work was to determine the level of benzene, toluene, o-xylene and m, p-xylene (BTX) in air samples collected from the cabins of new and used vehicles of the same model. Ten new vehicles were examined in order to check interior emission from materials used to equip the passenger compartment. In order to compare and define the impact of exhaust gases, air samples were also collected from two used cars, at different mileages (up to 20,000 km). All vehicles tested were of the same type. Samples were collected onto Carbograph 1TD sorbent, thermally desorbed and examined with the use of gas chromatography with flame ionisation and mass spectrometry detectors. All results obtained were referred to Polish and German requirements for indoor air quality (both in public buildings and in workspace environments). Average benzene, toluene, o-xylene and m, p-xylene concentrations in new cars were determined at the level of 11.8 microg/m3, 82.7 micro/m3, 21.2 microg/m3 and 89.5 micro/m3, respectively. In the used cars, BTX concentration increased with increasing vehicle mileage. The most significant increase of BTX concentration was observed above 11,000 km mileage.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%