Merck
  • Computer-controlled perifusion system for neuroendocrine tissues: development and applications.
Computer-controlled perifusion system for neuroendocrine tissues: development and applications.
Methods in enzymology (1986-01-01)
A Negro-Vilar, M D Culler
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Myoglobin from human heart, ≥95% (SDS-PAGE), buffered aqueous glycerol solution