Merck
  • Loss of MTUS1/ATIP expression is associated with adverse outcome in advanced bladder carcinomas: data from a retrospective study.
Loss of MTUS1/ATIP expression is associated with adverse outcome in advanced bladder carcinomas: data from a retrospective study.
BMC cancer (2014-03-22)
Anja Rogler, Sabine Hoja, Johannes Giedl, Arif B Ekici, Sven Wach, Helge Taubert, Peter J Goebell, Bernd Wullich, Michael Stöckle, Jan Lehmann, Sabrina Petsch, Arndt Hartmann, Robert Stoehr
ABSTRACT
Seventy percent of all bladder tumours tend to recur and need intensive surveillance, and a subset of tumours progress to muscle-invasive and metastatic disease. However, it is still difficult to find the adequate treatment for every individual patient as it is a very heterogeneous disease and reliable biomarkers are still missing. In our study we searched for new target genes in the critical chromosomal region 8p and investigated the potential tumour suppressor gene candidate MTUS1/ATIP in bladder cancer. MTUS1 was identified to be the most promising deleted target gene at 8p in aCGH analysis with 19 papillary bladder tumours. A correlation with bladder cancer was further validated using immunohistochemistry of 85 papillary and 236 advanced bladder tumours and in functional experiments. Kaplan-Meier analysis and multivariate Cox-regression addressed overall survival (OS) and disease-specific survival (DSS) as a function of MTUS1/ATIP expression. Bivariate correlations investigated associations between MTUS1/ATIP expression, patient characteristics and histopathology. MTUS1 expression was analysed in cell lines and overexpressed in RT112, where impact on viability, proliferation and migration was measured. MTUS1 protein expression was lost in almost 50% of all papillary and advanced bladder cancers. Survival, however, was only influenced in advanced carcinomas, where loss of MTUS1 was associated with adverse OS and DSS. In this cohort, there was also a significant correlation of MTUS1 expression and histological subtype: positive expression was detected in all micropapillary tumours and aberrant nuclear staining was detected in a subset of plasmocytoid urothelial carcinomas. MTUS1 was expressed in all investigated bladder cell lines and overexpression in RT112 led to significantly decreased viability. MTUS1 is a tumour suppressor gene in cultured bladder cancer cells and in advanced bladder tumours. It might represent one new target gene at chromosome 8p and can be used as an independent prognostic factor for advanced bladder cancer patients. The limitation of the study is the retrospective data analysis. Thus, findings should be validated with a prospective advanced bladder tumour cohort.
Sigma-Aldrich
BrdU Cell Proliferation Assay, This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid