Merck
  • Leucine modulates the effect of Walker factor, a proteolysis-inducing factor-like protein from Walker tumours, on gene expression and cellular activity in C2C12 myotubes.
Leucine modulates the effect of Walker factor, a proteolysis-inducing factor-like protein from Walker tumours, on gene expression and cellular activity in C2C12 myotubes.
Cytokine (2013-06-12)
Estela Maria Gonçalves, Emilianne Miguel Salomão, Maria Cristina Cintra Gomes-Marcondes
ABSTRACT
Cancer-cachexia causes severe weight loss, particularly from the wasting of skeletal muscle, which occurs due to increased protein catabolism and/or decreased protein synthesis. The muscle protein degradation observed in cancer patients is mediated by a specific cytokine, proteolysis-inducing factor (PIF), which is produced by the tumour. This protein increases the ubiquitin-proteasome pathway activity, and the synthesis of muscle protein in these patients can be affected by several factors, including nutrient-related signalling. Some nutrients, such as leucine, can decrease the ubiquitin-proteasome pathway activity and increase the skeletal muscle protein content in cachectic animals. In this study, we investigated the effects of leucine on cell viability, morphology, functional proteasome activity, enzymatic activity, and protein synthesis and degradation in C2C12 myotubes exposed to the proteolysis-inducing factor (PIF)-like protein purified from Walker tumour-bearing rats. Walker factor (WF) had no cytotoxic effects on myotube cells and morphological characteristics were not altered in the presence of WF and/or leucine. However, increased alkaline phosphatase activity was observed. At higher WF concentrations, chymotrypsin-like activity, cathepsin B activity and 20S proteasome gene expression increased. Treating myotubes with leucine before exposure to WF causes leads to a decrease in proteasome activity as well as the activity of chymotrypsin and cathepsin enzymes. Total protein synthesis decreased in WF-treated cells concomitantly as protein degradation increased. After leucine exposure, the observed effects of WF were minimal or even reverted in some cases. Taken together, these results suggest an important modulatory effect for leucine on the effects of WF in C2C12 myotube cells.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder