Merck
  • Comparative pharmacokinetics of subcutaneous trastuzumab administered via handheld syringe or proprietary single-use injection device in healthy males.
Comparative pharmacokinetics of subcutaneous trastuzumab administered via handheld syringe or proprietary single-use injection device in healthy males.
Cancer chemotherapy and pharmacology (2013-09-24)
Chris J Wynne, Rod B Ellis-Pegler, Devonie S Waaka, Christian Schwabe, Michaela Lehle, Dominik Heinzmann, Ranvir Mangat, Chunze Li, Anne E Dick, Nicola A Cranshaw, Bert L Lum
ABSTRACT
To demonstrate pharmacokinetic (PK) comparability for a single dose of 600 mg subcutaneous (SC) trastuzumab, administered via a novel single-use injection device (SID) or handheld syringe in 119 randomized healthy male subjects. The co-primary PK endpoints area under the time-concentration curve from the start of dosing to day 22 (AUC(0-21 days)) and maximum observed trastuzumab serum concentration (C(max)) were dose-normalized and body-weight-adjusted, and compared using geometric mean ratios (GMRs). SID performance, injection site pain, adverse events, and antidrug antibodies (ADAs) were assessed. GMRs and 90 % confidence intervals (CIs) were 1.01 (0.96-1.07) for AUC(0-21 days) and 1.02 (0.96-1.10) for C(max), which fell within the prespecified bioequivalence range (0.80-1.25). No SID quality issues or failures occurred. Adverse events were mostly mild, with no deaths, adverse event-related withdrawals, or life-threatening, cardiac, or serious events reported. The ADA rate was low, and no neutralizing antibodies were detected. Trastuzumab SC via SID demonstrated comparable PK and safety to handheld syringe administration. SID performance was very satisfactory.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid