Merck
  • A biomarker based detection and characterization of carcinomas exploiting two fundamental biophysical mechanisms in mammalian cells.
A biomarker based detection and characterization of carcinomas exploiting two fundamental biophysical mechanisms in mammalian cells.
BMC cancer (2013-12-07)
Martin Grimm, Steffen Schmitt, Peter Teriete, Thorsten Biegner, Arnulf Stenzl, Jörg Hennenlotter, Hans-Joachim Muhs, Adelheid Munz, Tatjana Nadtotschi, Klemens König, Jörg Sänger, Oliver Feyen, Heiko Hofmann, Siegmar Reinert, Johannes F Coy
ABSTRACT
Biomarkers allowing the characterization of malignancy and therapy response of oral squamous cell carcinomas (OSCC) or other types of carcinomas are still outstanding. The biochemical suicide molecule endonuclease DNaseX (DNaseI-like 1) has been used to identify the Apo10 protein epitope that marks tumor cells with abnormal apoptosis and proliferation. The transketolase-like protein 1 (TKTL1) represents the enzymatic basis for an anaerobic glucose metabolism even in the presence of oxygen (aerobic glycolysis/Warburg effect), which is concomitant with a more malignant phenotype due to invasive growth/metastasis and resistance to radical and apoptosis inducing therapies. Expression of Apo10 and TKTL1 was analysed retrospectively in OSCC specimen (n = 161) by immunohistochemistry. Both markers represent independent markers for poor survival. Furthermore Apo10 and TKTL1 have been used prospectively for epitope detection in monocytes (EDIM)-blood test in patients with OSCC (n = 50), breast cancer (n = 48), prostate cancer (n = 115), and blood donors/controls (n = 74). Positive Apo10 and TKTL1 expression were associated with recurrence of the tumor. Multivariate analysis demonstrated Apo10 and TKTL1 expression as an independent prognostic factor for reduced tumor-specific survival. Apo10+/TKTL1+ subgroup showed the worst disease-free survival rate in OSCC.EDIM-Apo10 and EDIM-TKTL1 blood tests allowed a sensitive and specific detection of patients with OSCC, breast cancer and prostate cancer before surgery and in after care. A combined score of Apo10+/TKTL1+ led to a sensitivity of 95.8% and a specificity of 97.3% for the detection of carcinomas independent of the tumor entity. The combined detection of two independent fundamental biophysical processes by the two biomarkers Apo10 and TKTL1 allows a sensitive and specific detection of neoplasia in a noninvasive and cost-effective way. Further prospective trials are warranted to validate this new concept for the diagnosis of neoplasia and tumor recurrence.
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
DNase I, Amplification Grade
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein