Merck
  • [The salivary factors related to caries and periodontal disease in children and adolescents with diabetes mellitus].
[The salivary factors related to caries and periodontal disease in children and adolescents with diabetes mellitus].
Zhonghua kou qiang yi xue za zhi = Zhonghua kouqiang yixue zazhi = Chinese journal of stomatology (2013-12-10)
Meng-xing Wang, Xin Wang, Zhi Zhang, Man Qin
ABSTRACT
To detect the salivary factors related to caries and periodontal disease and to analyze the risk of caries and periodontal disease in children and adolescents with diabetes mellitus. The study comprised 30 children with diabetic mellitus, aged 7-15 years old, and 60 healthy age-and gender-matched children. Caries and periodontal indexes were recorded and saliva related factors were analyzed. Caries indexes of diabetes children [permanent teeth: decay missing filling tooth (DMFT) M (Q1,Q3) = 0(0, 4), deciduous teeth: decay missing filling tooth (dmft) M (Q1,Q3) = 0(0, 1)] were not significantly different with those of healthy children [DMFT M (Q1,Q3) = 1(0, 3), dmft M (Q1,Q3) = 0(0, 4)], but plaque index (PLI) (1.25 ± 0.33) and bleeding index (BI) (0.74 ± 0.45) of diabetes children were significantly higher than those of healthy children (PLI was 0.93 ± 0.31,BI was 0.34 ± 0.22) (P < 0.001). Salivary pH of diabetes children (7.68 ± 0.36) was significantly higher than that of healthy children (7.30 ± 0.32) (P < 0.05), and salivary acid buffering capacity had no significant difference between the two groups (P > 0.05). Salivary glucose, immunoglobulin sIgA and sIgG were not significantly different between the two groups (P > 0.05).Salivary lysozyme of diabetes children was significantly higher than that of healthy children (P < 0.05). Total protein was significantly lower in diabetes children than in healthy children (P < 0.05). Salivary lactate dehydrogenase had no significant difference between the two groups (P > 0.05). Diabetes mellitus can lead to the changes of some salivary factors related to gingivitis in diabetes children. Children and adolescents with diabetes mellitus may have a higher risk of periodontal disease.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard