Merck
A novel influenza virus neuraminidase inhibitor AV5027.
Antiviral research (2014-01-15)
Alexandre V Ivachtchenko, Yan A Ivanenkov, Oleg D Mitkin, Pavel M Yamanushkin, Vadim V Bichko, Irina A Leneva, Olga V Borisova
ABSTRACT
A medium-sized focused library of novel Oseltamivir structural analogues with promising antiviral activity was successfully synthesized using a combinatorial approach. The synthesized compounds were then thoroughly evaluated in neuraminidase- and cell-based assays. As a result, (3R,4R,5S)-4-(2,2-difluoroacetylamino)-5-amino-3-(1-ethyl-propoxy)-cyclohex-1-enecarboxylic acid (AV5027) was identified as novel Hit-compound with picomolar potency. QSAR analysis was carried out based on the obtained biological data. Computational modeling was performed using a 3D-molecular docking approach and classical regression analysis. The developed integral model demonstrated a sufficient prediction accuracy and tolerance to evaluate compounds based on their potential activity against neuraminidase (NA) at least within the scaffold. Several compounds from the series can be reasonably regarded as promising anti-influenza drug-candidates.
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)