Merck
  • Long-term metabolic benefits of exenatide in mice are mediated solely via the known glucagon-like peptide 1 receptor.
Long-term metabolic benefits of exenatide in mice are mediated solely via the known glucagon-like peptide 1 receptor.
American journal of physiology. Regulatory, integrative and comparative physiology (2014-01-31)
Krystyna Tatarkiewicz, Emmanuel J Sablan, Clara J Polizzi, Christiane Villescaz, David G Parkes
ABSTRACT
Glucagon-like peptide 1 receptors (GLP-1R) are expressed in multiple tissues and activation results in metabolic benefits including enhanced insulin secretion, slowed gastric emptying, suppressed food intake, and improved hepatic steatosis. Limited and inconclusive knowledge exists regarding whether the effects of chronic exposure to a GLP-1R agonist are solely mediated via this receptor. Therefore, we examined 3-mo dosing of exenatide in mice lacking a functional GLP-1R (Glp1r(-/-)). Exenatide (30 nmol · kg(-1) · day(-1)) was infused subcutaneously for 12 wk in Glp1r(-/-) and wild-type (Glp1r(+/+)) control mice fed a high-fat diet. Glycated hemoglobin A1c (HbA1c), plasma glucose, insulin, amylase, lipase, alanine aminotransferase (ALT), aspartate aminotransferase (AST), body weight, food intake, terminal hepatic lipid content (HLC), and plasma exenatide levels were measured. At the end of the study, oral glucose tolerance test (OGTT) and rate of gastric emptying were assessed. Exenatide produced no significant changes in Glp1r(-/-) mice at study end. In contrast, exenatide decreased body weight, food intake, and glucose in Glp1r(+/+) mice. When compared with vehicle, exenatide reduced insulin, OGTT glucose AUC0-2h, ALT, and HLC in Glp1r(+/+) mice. Exenatide had no effect on plasma amylase or lipase levels. Exenatide concentrations were approximately eightfold higher in Glp1r(-/-) versus Glp1r(+/+) mice after 12 wk of infusion, whereas renal function was similar. These data support the concept that exenatide requires a functional GLP-1R to exert chronic metabolic effects in mice, and that novel "GLP-1" receptors may not substantially contribute to these changes. Differential exenatide plasma levels in Glp1r(+/+) versus Glp1r(-/-) mice suggest that GLP-1R may play an important role in plasma clearance of exenatide and potentially other GLP-1-related peptides.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g