Merck
  • Human recombinant lysozyme downregulates advanced glycation endproduct-induced interleukin-6 production and release in an in-vitro model of human proximal tubular epithelial cells.
Human recombinant lysozyme downregulates advanced glycation endproduct-induced interleukin-6 production and release in an in-vitro model of human proximal tubular epithelial cells.
Experimental biology and medicine (Maywood, N.J.) (2014-02-06)
Davide Gallo, Moreno Cocchietto, Elisa Masat, Chiara Agostinis, Elisa Harei, Paolo Veronesi, Gianni Sava
ABSTRACT
Diabetic nephropathy is the leading cause of chronic renal disease and one of the major causes of cardiovascular mortality. Evidence suggests that its progression is due to the chronic hyperglycemia consequent to the production and accumulation of advanced glycation endproducts (AGEs). Lysozyme was shown to posses AGE-sequestering properties and the capacity to reduce the severity of the early stage manifestations of the diabetic nephropathy. This study was aimed to contribute to the understanding the molecular mechanisms of lysozyme effectiveness in the diabetic nephropathy, using an in-vitro cellular model, represented by the HK-2 cells, human proximal tubular epithelial cells. Lysozyme significantly reduced the AGE-induced IL-6 mRNA and an ELISA assay showed also a decreased release of the functional protein with a dose-dependent trend. In addition, lysozyme prevented macrophage recruitment, suggesting its capacity to elicit an anti-inflammatory action. We may conclude that the protective action of lysozyme on the nephrotoxic effects of AGE may depend, at least in part, on its ability to prevent the production and release of inflammatory mediators, such as IL-6 and to reduce macrophage recruitment in the inflammatory sites.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material