Merck
  • Extended use of a selective inhibitor of acid lipase for the diagnosis of Wolman disease and cholesteryl ester storage disease.
Extended use of a selective inhibitor of acid lipase for the diagnosis of Wolman disease and cholesteryl ester storage disease.
Gene (2014-02-11)
G Civallero, J De Mari, C Bittar, M Burin, R Giugliani
ABSTRACT
Lysosomal acid lipase (LAL) deficiency produces two well defined inborn disorders, Wolman disease (WD) and cholesteryl ester storage disease (CESD). WD is a severe, early-onset condition involving massive storage of triglycerides and cholesteryl esters in the liver, with death usually occurring before one year of life. CESD is a more attenuated, later-onset disease that leads to a progressive and variable liver dysfunction. Diagnosis of LAL deficiency is mainly based on the enzyme assay of LAL activity in fibroblasts. Recently, a selective acid lipase inhibitor was used for the determination of enzyme activity in dried-blood filter paper (DBFP) samples. To extend and to validate these studies, we tested LAL activity with selective inhibition on DBFP samples, leukocytes and fibroblasts. Our results showed a clear discrimination between patients with LAL deficiency and healthy controls when using DBFP, leukocytes or fibroblasts (p<0.001). Deficiency of LAL was also demonstrated in individuals referred to our laboratory with suspected clinical diagnosis of WD, CESD, and Niemann-Pick type B. We conclude that the assay of LAL using selective inhibitor is a reliable and useful method for the identification of LAL deficiency, not only in DBFP samples but also in leukocytes and fibroblasts. This is important as enzyme replacement therapy for LAL deficiency is currently being developed, making the correct diagnosis a critical issue.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas fluorescens, lyophilized powder, ≥10,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas sp., lyophilized powder, ≥200,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from bovine pancreas, lyophilized powder, ≥200 units/g protein
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g