Merck
  • Cell cycle characteristics of the pancreas in an animal model of isolated pancreatic trauma.
Cell cycle characteristics of the pancreas in an animal model of isolated pancreatic trauma.
The journal of trauma and acute care surgery (2014-02-21)
Dai Rui-Wu, Chen Guang-Yu, He Fa-Qun, Huang Zu, Yan Hong-Tao, Liang Hong-Yin, Wang Tao, Lin Ning, Tang Li-Jun, Chen Li-Ping
ABSTRACT
In our previous study, we established a small animal model that mimicked the pathophysiology of isolated pancreatic trauma. To gain further insights into the relationships between tissue damage and the ability of the pancreatic cells to regenerate, we induced pancreatic trauma in rats maintained over 7 days and analyzed both the alteration of the cell death and the cell cycle distribution of the pancreatic cells in this study. The rats were divided into two groups as follows: impact and control. The pancreas in the impact group was injured by a BIM-III biotical impact machine. Pancreatic enzyme activity, the level of Ca in the serum, pancreatic cell death, and cell cycle characteristics were examined after the trauma. In the impact groups, lipase was activated later than amylase and lasted persistently. The levels of serum Ca decreased at 6 hours after injury, sharply declined at 24 hours and 72 hours compared with the control groups, and returned to normal levels at 7 days. The pancreatic trauma also induced the compensatory proliferation of pancreatic cells. The results from a TUNEL stain, flow cytometry, Western blot, and immunohistochemistry indicated that pancreatic trauma induces cell death and the compensatory proliferation of pancreatic cells. Detecting amylase and lipase at the same time can help us determine the exocrine function of pancreas. Serum Ca can be used as an indicator for estimating the severity of pancreatic trauma. The cell cycle characteristics of the pancreas in the animal model of isolated pancreatic trauma indicate that the proper remedial time is in the first 24 hours after the pancreatic trauma.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g