Merck
  • An RNA aptamer possessing a novel monovalent cation-mediated fold inhibits lysozyme catalysis by inhibiting the binding of long natural substrates.
An RNA aptamer possessing a novel monovalent cation-mediated fold inhibits lysozyme catalysis by inhibiting the binding of long natural substrates.
RNA (New York, N.Y.) (2014-02-27)
Camille S Padlan, Vladimir N Malashkevich, Steve C Almo, Matthew Levy, Michael Brenowitz, Mark E Girvin
ABSTRACT
RNA aptamers are being developed as inhibitors of macromolecular and cellular function, diagnostic tools, and potential therapeutics. Our understanding of the physical nature of this emerging class of nucleic acid-protein complexes is limited; few atomic resolution structures have been reported for aptamers bound to their protein target. Guided by chemical mapping, we systematically minimized an RNA aptamer (Lys1) selected against hen egg white lysozyme. The resultant 59-nucleotide compact aptamer (Lys1.2minE) retains nanomolar binding affinity and the ability to inhibit lysozyme's catalytic activity. Our 2.0-Å crystal structure of the aptamer-protein complex reveals a helical stem stabilizing two loops to form a protein binding platform that binds lysozyme distal to the catalytic cleft. This structure along with complementary solution analyses illuminate a novel protein-nucleic acid interface; (1) only 410 Å(2) of solvent accessible surface are buried by aptamer binding; (2) an unusually small fraction (∼18%) of the RNA-protein interaction is electrostatic, consistent with the limited protein phosphate backbone contacts observed in the structure; (3) a single Na(+) stabilizes the loops that constitute the protein-binding platform, and consistent with this observation, Lys1.2minE-lysozyme complex formation takes up rather than displaces cations at low ionic strength; (4) Lys1.2minE inhibits catalysis of large cell wall substrates but not catalysis of small model substrates; and (5) the helical stem of Lys1.2minE can be shortened to four base pairs (Lys1.2minF) without compromising binding affinity, yielding a 45-nucleotide aptamer whose structure may be an adaptable protein binding platform.
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)