Merck
  • The antioxidant effects of pumpkin seed oil on subacute aflatoxin poisoning in mice.
The antioxidant effects of pumpkin seed oil on subacute aflatoxin poisoning in mice.
Environmental toxicology (2014-03-05)
Gökhan Eraslan, Murat Kanbur, Öznur Aslan, Mürsel Karabacak
ABSTRACT
This study was aimed at the investigation of the antioxidant effect of pumpkin seed oil against the oxidative stress-inducing potential of aflatoxin. For this purpose, 48 male BALB/c mice were used. Four groups, each comprising 12 mice, were established. Group 1 was maintained as the control group. Group 2 was administered with pumpkin seed oil alone at a dose of 1.5 mL/kg.bw/day (∼1375mg/kg.bw/day). Group 3 received aflatoxin (82.45% AFB1 , 10.65% AFB2 , 4.13% AFG1, and 2.77% AFG2 ) alone at a dose of 625 μg/kg.bw/day. Finally, group 4 was given both 1.5 mL/kg.bw/day pumpkin seed oil and 625 μg/kg.bw/day aflatoxin. All administrations were oral, performed with the aid of a gastric tube and continued for a period of 21 days. At the end of day 21, the liver, lungs, kidneys, brain, heart, and spleen of the animals were excised, and the extirpated tissues were homogenized appropriately. Malondialdehyde (MDA) levels and catalase (CAT), superoxide dismutase (SOD), and glutathione peroxidase (GSH-Px) activities were determined in tissue homogenates. In conclusion, it was determined that aflatoxin exhibited adverse effects on most of the oxidative stress markers. The administration of pumpkin seed oil diminished aflatoxin-induced adverse effects. In other words, the values of the group, which was administered with both aflatoxin and pumpkin seed oil, were observed to have drawn closer to the values of the control group.
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder