Merck
  • Developments in x-ray crystallographic structure determination of biological macromolecules.
Developments in x-ray crystallographic structure determination of biological macromolecules.
Science (New York, N.Y.) (2014-03-08)
Elspeth F Garman
ABSTRACT
The three-dimensional structures of large biomolecules important in the function and mechanistic pathways of all living systems and viruses can be determined by x-ray diffraction from crystals of these molecules and their complexes. This area of crystallography is continually expanding and evolving, and the introduction of new methods that use the latest technology is allowing the elucidation of ever larger and more complex biological systems, which are now becoming tractable to structure solution. This review looks back at what has been achieved and forward at how current and future developments may allow technical challenges to be overcome.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein