Merck
  • Interaction of multi-walled carbon nanotubes with water-soluble proteins: effect of sidewall carboxylation.
Interaction of multi-walled carbon nanotubes with water-soluble proteins: effect of sidewall carboxylation.
Journal of nanoscience and nanotechnology (2014-04-17)
Tomoya Takada, Rei Kurosaki, Yuji Konno, Shigeaki Abe
ABSTRACT
Effect of sidewall carboxylation on protein adsorption behavior of multi-walled carbon nanotubes (MWCNTs) was studied. Two water-soluble proteins, bovine serum albumin (BSA) and egg white lysozyme (LYS), were employed in this work. Carboxylation of MWCNTs suppressed adsorption of BSA, whereas adsorption of LYS was enhanced by the carboxylation. These behaviors are explained by the difference in the dominance of hydrophobic interaction and ionic interaction between MWCNTs and the proteins.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard