Merck
  • Single center experience with total body irradiation and melphalan (TBI-MEL) myeloablative conditioning regimen for allogeneic stem cell transplantation (SCT) in patients with refractory hematologic malignancies.
Single center experience with total body irradiation and melphalan (TBI-MEL) myeloablative conditioning regimen for allogeneic stem cell transplantation (SCT) in patients with refractory hematologic malignancies.
Annals of hematology (2013-10-08)
Bhavana Bhatnagar, Aaron P Rapoport, Hong-Bin Fang, Can Ilyas, Deniz Marangoz, Vinil Akbulut, Kathleen Ruehle, Ashraf Badros, Saul Yanovich, Görgün Akpek
ABSTRACT
We retrospectively evaluated the tolerability and efficacy of fractionated total body irradiation (TBI) (1,200 cGy) and melphalan (MEL) (100-110 mg/m(2)) myeloablative conditioning in 48 patients with nonremission AML (n = 14), ALL (n = 10), NHL (n = 18), and other refractory hematologic malignancies (n = 6) who received allogeneic stem cell transplantation (SCT) between 2002 and 2011. Median age was 48 years (22 to 68); 14 out of 26 leukemia patients (54 %) had circulating blasts at transplant, 20 (50 %) evaluable patients had poor-risk cytogenetics, 12 (25 %) had prior SCT, and 10 (21 %) received stem cells from a mismatch donor. All patients received tacrolimus with or without methotrexate for GVHD prophylaxis. At the time of analysis, 13 patients (27 %) were alive and disease free. Engraftment was complete in all patients. The median time to ANC recovery (>500) was 12 days (range, 6-28). The most common grade III and IV toxicities were mucositis and infections. Eighteen patients (43 %) developed grade II-IV acute GVHD, and eight (26 %) had extensive chronic GVHD. Of 44 evaluable patients for response, 28 (64 %) achieved a complete remission (CR), and seven (15 %) had a partial remission after the transplant. With a median follow-up of 30 months (4 to 124 months) for surviving patients, the cumulative incidence of relapse was 45 % at 1 year, and the probability of overall survival (OS) at 5 years was 22.5 %. Multivariate analysis showed that platelet count (<80,000/mL) and lactic dehydrogenase (>500 IU/L) at SCT were associated with relapse. Age less than 53 years and CR after SCT were associated with better OS. Our data suggest that TBI-MEL can result in CR in two thirds, durable remission in one third, and 5-year survival in about one quarter of patients with nonremission hematologic malignancies. Further studies with TBI-MEL in standard risk transplant patients are warranted.
Sigma-Aldrich
Melphalan, powder