Merck
  • Anti-carbamylated protein (anti-CarP) antibodies precede the onset of rheumatoid arthritis.
Anti-carbamylated protein (anti-CarP) antibodies precede the onset of rheumatoid arthritis.
Annals of the rheumatic diseases (2013-12-18)
Jing Shi, Lotte A van de Stadt, E W Nivine Levarht, Tom W J Huizinga, Dörte Hamann, Dirkjan van Schaardenburg, René E M Toes, Leendert A Trouw
ABSTRACT
The presence of anti-citrullinated protein antibodies (ACPA) and IgM-rheumatoid factor (IgM-RF) years before the clinical diagnosis of rheumatoid arthritis (RA) suggests they are possibly involved in the pathogenic process underlying RA. In this study, we analysed whether anti-carbamylated protein (anti-CarP) antibodies, a novel autoantibody system against carbamylated proteins, can also be detected in healthy individuals before they developed RA. Multiple sera from asymptomatic blood donors prior to the onset of their RA symptoms and sera from age-matched and sex-matched controls were tested for the presence of antibodies directed against carbamylated-fetal calf serum (Ca-FCS), carbamylated-fibrinogen (Ca-Fib), cyclic citrullinated-peptide 2 and IgM-RF. Anti-Ca-FCS and anti-Ca-Fib antibodies were each present in 27% and 38% of the last serum samples of blood donors prior to the diagnosis of RA. Both anti-Ca-FCS and anti-Ca-Fib antibodies could be detected many years before the onset of RA. Anti-CarP antibodies as well as ACPA are, on average, detected earlier than IgM-RF. In addition to ACPA and IgM-RF, also the newly identified anti-CarP antibodies appear many years before the diagnosis of RA.
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)