Merck
  • Kanamycin-furosemide ototoxicity in the mouse cochlea: a 3-dimensional analysis.
Kanamycin-furosemide ototoxicity in the mouse cochlea: a 3-dimensional analysis.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2014-01-15)
Heather M Schmitz, Shane B Johnson, Peter A Santi
ABSTRACT
Administration of an aminoglycoside antibiotic and loop diuretic causes damage to hair cells in the organ of Corti, resulting in their death and the death of their corresponding spiral ganglion neurons. While this phenomenon has been studied previously, analysis of its effects in the whole cochlea has not been reported. The authors sought to evaluate the effects of a combination dose of kanamycin and furosemide in mice cochlea using an imaging system and computer analysis that allowed for nondestructive, whole-cochlea visualization. Study using an animal model. Cochlear analysis laboratory. Five mice received kanamycin and furosemide and 3 mice received saline. Cochleas were harvested and imaged with scanning thin-sheet laser imaging microscopy (sTSLIM) to analyze sensory cells and cochlea structures. The drug-treated animals showed substantial loss of inner hair cells and complete outer hair cell loss. All treated mice showed spiral ganglion neuron loss with fewer neurons than control animals and decreased cell density in the middle turn of the cochlea. The spiral ligament and spiral limbus in the treated animals also showed a decrease in fibrocyte cell density in the middle to apical portion of the cochlea. The stria vascularis appeared normal in all animals. Imaging methods that allow for whole-cochlea analysis provide insight into changes that occur in the cochlea after ototoxic insult. Trends that may not be apparent in cross-section samples of the cochlea can be observed. Computer analysis of these trends allows them to be assessed accurately.
Product Description
Furosemide for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Furosemide