Merck
  • Cladribine, cytarabine, filgrastim, and mitoxantrone (CLAG-M) compared to standard induction in acute myeloid leukemia from myelodysplastic syndrome after azanucleoside failure.
Cladribine, cytarabine, filgrastim, and mitoxantrone (CLAG-M) compared to standard induction in acute myeloid leukemia from myelodysplastic syndrome after azanucleoside failure.
Leukemia research (2014-01-21)
Michael V Jaglal, Vu H Duong, Celeste M Bello, Najla H Al Ali, Eric Padron, Hugo F Fernandez, Alan F List, Jeffrey E Lancet, Rami S Komrokji
ABSTRACT
For patients with acute myeloid leukemia from antecedent myelodysplastic syndrome particularly after azanucleoside treatment failure, outcome is poor. Here, we conducted a case-control study in these patients to compare the efficacy of CLAG-M induction (28 patients) versus standard 3+7 induction chemotherapy (24 patients). Response rates (P=0.014) and median overall survival (P=0.025) were 64% and 202 days (95% CI 37-367 days) versus 29% and 86 days (95% CI 36-136) in the CLAG-M and 3+7 cohorts, respectively. Median overall survival was 202 (95% CI 37-367 days) versus 86 days (95% CI 36-136) (P=0.025), respectively. CLAG-M has encouraging activity in this patient group.
Product Description
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Mitoxantrone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
2-Chloro-2′-deoxyadenosine, antileukemic
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Cladribine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cladribine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Filgrastim, European Pharmacopoeia (EP) Reference Standard