Merck
  • Does dexamethasone induce more neuropsychological side effects than prednisone in pediatric acute lymphoblastic leukemia? A systematic review.
Does dexamethasone induce more neuropsychological side effects than prednisone in pediatric acute lymphoblastic leukemia? A systematic review.
Pediatric blood & cancer (2014-02-18)
L T Warris, M M van den Heuvel-Eibrink, M A H den Hoed, F K Aarsen, R Pieters, E L T van den Akker
ABSTRACT
Steroid-induced neuropsychological side effects impact quality of life in children with acute lymphoblastic leukemia. Dexamethasone induces more metabolic side effects than prednisone. To evaluate whether dexamethasone also leads to more neuropsychological side effects, we reviewed all available literature. Randomized controlled trials with neuropsychological function as the primary or secondary outcome did not show clinically meaningful differences between dexamethasone and prednisone on cognition, mood or behavior.
Product Description
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material