Merck
  • The relation between hairpin formation by mitochondrial WANCY tRNAs and the occurrence of the light strand replication origin in Lepidosauria.
The relation between hairpin formation by mitochondrial WANCY tRNAs and the occurrence of the light strand replication origin in Lepidosauria.
Gene (2014-02-25)
Hervé Seligmann, Antonieta Labra
ABSTRACT
Mitochondrial light strand DNA replication is initiated at light strand replication origins (OLs), short stem-loop hairpins formed by the heavy strand DNA. OL-like secondary structures are also formed by heavy strand DNA templating for the five tRNAs adjacent to OLs, the WANCY tRNA cluster. We tested whether natural OL absence associates with greater capacities for formation of OL-like structures by WANCY tRNA genes. Using lepidosaurian taxa (Sphenodon, lizards and amphisbaenids), we compared WANCY tRNA capacities to form OL-like structures between 248 taxa possessing an OL with 131 taxa without OL (from different families). On average, WANCY tRNA genes form more OL-like structures in the absence of a regular OL than in its presence. Formation of OL-like structures by WANCY tRNAs follows hierarchical patterns that may reduce competition between the tRNA's translational function and its secondary OL function: the rarer the tRNA's cognate amino acid, the greater the capacity to form OL-like structures. High OL-forming capacities for neighboring tRNAs are avoided. Because OL absence usually occurs in taxa with reduced genomes, increased formation of OL-like structures by WANCY tRNAs might result from selection for greater metabolic efficiency. Further analyses suggest that OL loss is one of the latest steps in genome reduction, and promotes the increase in formation of OL-like structures by WANCY tRNA genes in Lepidosauria.
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid