Merck
  • Influence of parafascicular thalamic input on neuronal activity within the nucleus accumbens is mediated by nitric oxide - an in vivo study.
Influence of parafascicular thalamic input on neuronal activity within the nucleus accumbens is mediated by nitric oxide - an in vivo study.
Life sciences (2014-03-13)
Michaela M Kraus, Helmut Prast, Athineos Philippu
ABSTRACT
Thalamostriatal fibers are involved in cognitive tasks such as acquisition, learning, processing of sensory events, and behavioral flexibility and might play a role in Parkinson's disease. The aim of the present study was the in vivo electrochemical characterization of the projection from the lateral aspect of the parafascicular thalamus (Pfl) to the dorsolateral aspect of the nucleus accumbens (dNAc). Since nitric oxide (NO) plays a crucial role in striatal synaptic transmission, its implication in Pfl-evoked signaling within the dNAc was investigated. The Pfl was electrically stimulated utilizing paired pulses and extracellular potentials were recorded within the dNAc. Simultaneously, the dNAc was superfused using the push-pull superfusion technique for local application of compounds and for assessing the influence of NO on release of glutamate, aspartate and GABA. Stimulation of the Pfl evoked a negative-going component at 9-14 ms followed by a positive-going component at 39-48 ms. The early response was current-dependent and diminished by superfusion of the dNAc with tetrodotoxin, kynurenic acid or N(G)-nitro-l-arginine methyl ester (L-NAME), while 3-(2-hydroxy-2-nitroso-1-propylhydrazino)-1-propanamine (PAPA/NO) increased this evoked potential. Transmitter release was inhibited by L-NAME and facilitated by PAPA/NO. This study describes for the first time in vivo extracellular electrical responses of the dNAc on stimulation of the Pfl. Synaptic transmission within the dNAc on stimulation of the Pfl seems to be facilitated by NO.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamic acid, FCC
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland